5-Chlor-1,3-benzodioxol

Lieferant: FLUOROCHEM

Synonyme: 5-Chlorobenzo[d][1,3]dioxole

018400-5G
FLUO018400-5GEA 69.8 EUR
FLUO018400-5G
5-Chlor-1,3-benzodioxol
5-Chlor-1,3-benzodioxol
Formel: C₇H₅ClO₂
Molecular Weight: 156,57 g/mol
MDL: MFCD00010842
CAS-Nummer: 7228-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR