3-Chlor-o-kresol

Lieferant: FLUOROCHEM

018406-10G
FLUO018406-10GEA 104 EUR
FLUO018406-10G
3-Chlor-o-kresol
3-Chlor-o-kresol
Formel: C₇H₇ClO
MDL: MFCD00079738
CAS-Nummer: 3260-87-5
EINECS: 221-861-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR