2-Chlor-4-nitroanisol

Lieferant: FLUOROCHEM

Synonyme: 1-Chloro-2-methoxy-5-nitrobenzene

018415-10G
FLUO018415-10GEA 0 EUR
FLUO018415-10G
2-Chlor-4-nitroanisol
2-Chlor-4-nitroanisol
Formel: C₇H₆ClNO₃
MDL: MFCD00060681
CAS-Nummer: 4920-79-0
EINECS: 225-547-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR