2-Chlor-4-nitrobenzamid
Lieferant: FLUOROCHEM
Synonyme:
Aklomid
018416-10G
FLUO018416-10GEA
234
EUR
FLUO018416-10G
2-Chlor-4-nitrobenzamid
2-Chlor-4-nitrobenzamid
Formel:
C₇H₅ClN₂O₃ |
MDL:
MFCD00017119 CAS-Nummer: 3011-89-0 EINECS: 221-143-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...