2-Chlor-4-nitrobenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-4-nitrobenzol-1-carbonylchlorid, 2-Chlor-4-nitrobenzoesäurechlorid
018418-10G
FLUO018418-10GEA
107
EUR
FLUO018418-10G
2-Chlor-4-nitrobenzoylchlorid
2-Chlor-4-nitrobenzoylchlorid
Formel:
C₇H₃Cl₂NO₃ Molecular Weight: 220,01 g/mol Siedepunkt: 112…116 °C (0,35 mmHg) Schmelzpunkt: 115 °C |
MDL:
MFCD00051515 CAS-Nummer: 7073-36-1 EINECS: 230-367-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...