2-Chlor-5-nitrobenzoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-5-nitrobenzol-1-carbonylchlorid, 2-Chlor-5-nitrobenzoesäurechlorid

018419-10G
FLUO018419-10GEA 0 EUR
FLUO018419-10G
2-Chlor-5-nitrobenzoylchlorid
2-Chlor-5-nitrobenzoylchlorid
Formel: C₇H₃Cl₂NO₃
Molecular Weight: 220,01 g/mol
Siedepunkt: 157…158 °C (11 mmHg)
Schmelzpunkt: 60 °C
MDL: MFCD00059180
CAS-Nummer: 25784-91-2
EINECS: 247-262-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR