2-(4-Chlorphenyl)ethylisothiocyanat
Lieferant: FLUOROCHEM
018422-1G
FLUO018422-1GEA
76.2
EUR
FLUO018422-1G
2-(4-Chlorphenyl)ethylisothiocyanat
2-(4-Chlorphenyl)ethylisothiocyanat
Formel:
C₉H₈ClNS Molecular Weight: 197,69 g/mol |
MDL:
MFCD00041127 CAS-Nummer: 17608-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...