1-(2-Chlorphenyl)imidazol

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Chlorphenyl)-1H-imidazol

018425-1G
FLUO018425-1GEA 195 EUR
FLUO018425-1G
1-(2-Chlorphenyl)imidazol
1-(2-Chlorophenyl)imidazole
Formel: C₉H₇ClN₂
MDL: MFCD00041205
CAS-Nummer: 51581-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR