1-(3-Chlorphenyl)imidazol

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Chlorphenyl)-1H-imidazol

018426-1G
FLUO018426-1GEA 195 EUR
FLUO018426-1G
1-(3-Chlorphenyl)imidazol
1-(3-Chlorophenyl)imidazole
Formel: C₉H₇ClN₂
MDL: MFCD00041207
CAS-Nummer: 51581-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR