1-(4-Chlorphenyl)imidazol

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Chlorphenyl)-1H-imidazol

018427-1G
FLUO018427-1GEA 152 EUR
FLUO018427-1G
1-(4-Chlorphenyl)imidazol
1-(4-Chlorophenyl)imidazole
Formel: C₉H₇ClN₂
MDL: MFCD00041208
CAS-Nummer: 51581-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR