3-(2-Chlorphenyl)propionsäure

Lieferant: FLUOROCHEM

018433-5G
FLUO018433-5GEA 67.1 EUR
FLUO018433-5G
3-(2-Chlorphenyl)propionsäure
3-(2-Chlorphenyl)propionsäure
Formel: C₉H₉ClO₂
MDL: MFCD00016547
CAS-Nummer: 1643-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR