3-(4-Chlorphenyl)propiononitril

Lieferant: FLUOROCHEM

Synonyme: 3-(4-Chlorophenyl)propanenitrile

018436-1G
FLUO018436-1GEA 139 EUR
FLUO018436-1G
3-(4-Chlorphenyl)propiononitril
3-(4-Chlorphenyl)propiononitril
Formel: C₉H₈ClN
Molecular Weight: 165,6195 g/mol
MDL: MFCD01310802
CAS-Nummer: 32327-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR