4-(2-Chlorphenyl)-3-thiosemicarbazid

Lieferant: FLUOROCHEM

018444-5G
FLUO018444-5GEA 224 EUR
FLUO018444-5G
4-(2-Chlorphenyl)-3-thiosemicarbazid
4-(2-Chlorphenyl)-3-thiosemicarbazid
Formel: C₇H₈ClN₃S
MDL: MFCD00041271
CAS-Nummer: 42135-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR