4-(3-Chlorphenyl)-3-thiosemicarbazid

Lieferant: FLUOROCHEM

018445-5G
FLUO018445-5GEA 177 EUR
FLUO018445-5G
4-(3-Chlorphenyl)-3-thiosemicarbazid
4-(3-Chlorphenyl)-3-thiosemicarbazid
Formel: C₇H₈ClN₃S
Molecular Weight: 201,68 g/mol
MDL: MFCD00041287
CAS-Nummer: 42135-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR