4-(4-Chlorphenyl)-3-thiosemicarbazid
Lieferant: FLUOROCHEM
Synonyme:
N-(4-Chlorophenyl)hydrazinecarbothioamide
018446-5G
FLUO018446-5GEA
195
EUR
FLUO018446-5G
4-(4-Chlorphenyl)-3-thiosemicarbazid
4-(4-Chlorphenyl)-3-thiosemicarbazid
Formel:
C₇H₈ClN₃S Molecular Weight: 201,68 g/mol |
MDL:
MFCD00041295 CAS-Nummer: 22814-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...