4-Chlorphenylharnstoff

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Chlorophenyl)urea

018451-10G
FLUO018451-10GEA 137 EUR
FLUO018451-10G
4-Chlorphenylharnstoff
4-Chlorphenylharnstoff
Formel: C₇H₇ClN₂O
Molecular Weight: 170,6 g/mol
MDL: MFCD00014788
CAS-Nummer: 140-38-5
EINECS: 205-412-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR