6-Chlorchinaldin

Lieferant: FLUOROCHEM

018454-10G
FLUO018454-10GEA 0 EUR
FLUO018454-10G
6-Chlorchinaldin
6-Chlorchinaldin
Formel: C₁₀H₈ClN
Molecular Weight: 177.63 g/mol
MDL: MFCD00051735
CAS-Nummer: 92-46-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR