8-Chlorchinaldin

Lieferant: FLUOROCHEM

Synonyme: 8-Chloro-2-methylquinoline

018455-10G
FLUO018455-10GEA 432 EUR
FLUO018455-10G
8-Chlorchinaldin
8-Chlorchinaldin
Formel: C₁₀H₈ClN
Molecular Weight: 177.63 g/mol
MDL: MFCD00051764
CAS-Nummer: 3033-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR