4-Chlor-m-xylol
Lieferant: FLUOROCHEM
|
Achtung
|
018462-5G
FLUO018462-5GEA
76.2
EUR
FLUO018462-5G
4-Chlor-m-xylol
4-Chlor-m-xylol
Formel:
C₈H₉Cl Molecular Weight: 140.61 g/mol |
MDL:
MFCD00060644 CAS-Nummer: 95-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...