2-Hydroxy-6-methylnicotinonitril

Lieferant: FLUOROCHEM

Synonyme: 6-Methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-Cyano-6-methyl-2-pyridone

018469-10G
FLUO018469-10GEA 90.9 EUR
FLUO018469-10G
2-Hydroxy-6-methylnicotinonitril
2-Hydroxy-6-methylnicotinonitril
Formel: C₇H₆N₂O
Molecular Weight: 134,14 g/mol
MDL: MFCD00006013
CAS-Nummer: 4241-27-4
EINECS: 224-202-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR