1-(3-Cyanphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

018471-1G
FLUO018471-1GEA 260 EUR
FLUO018471-1G
1-(3-Cyanphenyl)-2-thioharnstoff
1-(3-Cyanphenyl)-2-thioharnstoff
Formel: C₈H₇N₃S
MDL: MFCD00060439
CAS-Nummer: 41835-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR