1-(4-Cyanphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: N-(4-Cyanophenyl)thiourea

018472-1G
FLUO018472-1GEA 98.4 EUR
FLUO018472-1G
1-(4-Cyanphenyl)-2-thioharnstoff
1-(4-Cyanphenyl)-2-thioharnstoff
Formel: C₈H₇N₃S
Molecular Weight: 177,23 g/mol
MDL: MFCD00041175
CAS-Nummer: 3460-55-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR