3-(Cyclohexylamino)propionitril

Lieferant: FLUOROCHEM

018475-10G
FLUO018475-10GEA 0 EUR
FLUO018475-10G
3-(Cyclohexylamino)propionitril
3-(Cyclohexylamino)propionitril
Formel: C₉H₁₆N₂
MDL: MFCD00021325
CAS-Nummer: 702-03-4
EINECS: 211-864-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR