5-Cyclohexyl-o-anisidin

Lieferant: FLUOROCHEM

018476-1G
FLUO018476-1GEA 195 EUR
FLUO018476-1G
5-Cyclohexyl-o-anisidin
5-Cyclohexyl-o-anisidin
Formel: C₁₃H₁₉NO
MDL: MFCD00270102
CAS-Nummer: 206559-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR