4-Cyclohexyl-3-thiosemicarbazid
Lieferant: FLUOROCHEM
018479-10G
FLUO018479-10GEA
312
EUR
FLUO018479-10G
4-Cyclohexyl-3-thiosemicarbazid
4-Cyclohexyl-3-thiosemicarbazid
Formel:
C₇H₁₅N₃S |
MDL:
MFCD00021318 CAS-Nummer: 21198-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...