3-(Cyclopentylamino)propionitril

Lieferant: FLUOROCHEM

018483-10G
FLUO018483-10GEA 0 EUR
FLUO018483-10G
3-(Cyclopentylamino)propionitril
3-(Cyclopentylamino)propionitril
Formel: C₈H₁₄N₂
MDL: MFCD00060286
CAS-Nummer: 1074-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR