4,4'-Diacetylbiphenyl

Lieferant: FLUOROCHEM

018491-5G
FLUO018491-5GEA 0 EUR
FLUO018491-5G
4,4'-Diacetylbiphenyl
4,4'-Diacetylbiphenyl
Formel: C₁₆H₁₄O₂
Molecular Weight: 238.29 g/mol
MDL: MFCD00017248
CAS-Nummer: 787-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR