2,5-Dibrombenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2,5-Dibrombenzolsulfochlorid, 2,5-Dibrombenzolsulfonsäurechlorid, 2,5-Dibrombenzol-1-sulfonylchlorid

018497-5G
FLUO018497-5GEA 58.1 EUR
FLUO018497-5G
2,5-Dibrombenzolsulfonylchlorid
2,5-Dibrombenzolsulfonylchlorid
Formel: C₆H₃Br₂ClO₂S
Molecular Weight: 334,42 g/mol
MDL: MFCD00060685
CAS-Nummer: 23886-64-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR