3,5-Dibrom-4-hydroxybenzoesäure
Lieferant: FLUOROCHEM
|
Achtung
|
018500-10G
FLUO018500-10GEA
0
EUR
FLUO018500-10G
3,5-Dibrom-4-hydroxybenzoesäure
3,5-Dibromo-4-hydroxybenzoic acid
Formel:
C₇H₄Br₂O₃ Molecular Weight: 295,91 g/mol Siedepunkt: 365 °C (1013 hPa) Schmelzpunkt: 275…276 °C |
MDL:
MFCD00002548 CAS-Nummer: 3337-62-0 EINECS: 222-075-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...