2,3-Dibrompropylisothiocyanat

Lieferant: FLUOROCHEM

018504-10G
FLUO018504-10GEA 377 EUR
FLUO018504-10G
2,3-Dibrompropylisothiocyanat
2,3-Dibrompropylisothiocyanat
Formel: C₄H₅Br₂NS
MDL: MFCD00060411
CAS-Nummer: 51784-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR