2,5-Dibrom-m-xylol
Lieferant: FLUOROCHEM
Synonyme:
2,5-Dibrom-1,3-dimethylbenzol, 1,4-Dibrom-2,6-dimethylbenzol
018507-5G
FLUO018507-5GEA
72.9
EUR
FLUO018507-5G
2,5-Dibrom-m-xylol
2,5-Dibrom-m-xylol
Formel:
C₈H₈Br₂ Molecular Weight: 263,96 g/mol |
MDL:
MFCD00079745 CAS-Nummer: 100189-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...