N,N'-Bis(tert-butyl)ethylendiamin

Lieferant: FLUOROCHEM

018508-10G
FLUO018508-10GEA 0 EUR
FLUO018508-10G
N,N'-Bis(tert-butyl)ethylendiamin
N,N'-Bis(tert-butyl)ethylendiamin
Formel: C₁₀H₂₄N₂
MDL: MFCD00014996
CAS-Nummer: 4062-60-6
EINECS: 223-769-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR