1,3-Dibutyl-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: N,N'-Dibutylthioharnstoff

018509-50G
FLUO018509-50GEA 0 EUR
FLUO018509-50G
1,3-Dibutyl-2-thioharnstoff
1,3-Dibutyl-2-thioharnstoff
Formel: H₃CCH₂CH₂CH₂NHC(S)NHCH₂CH₂CH₂CH₃
Molecular Weight: 188,34 g/mol
Siedepunkt: 252 °C (1013 hPa)
Schmelzpunkt: 62…65 °C
MDL: MFCD00004926
CAS-Nummer: 109-46-6
EINECS: 203-674-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR