3,4-Dichlorbenzonitril

Lieferant: FLUOROCHEM

018515-5G
FLUO018515-5GEA 0 EUR
FLUO018515-5G
3,4-Dichlorbenzonitril
3,4-Dichlorbenzonitril
Formel: C₇H₃Cl₂N
Molecular Weight: 172,01 g/mol
Schmelzpunkt: 86…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016379
CAS-Nummer: 6574-99-8
EINECS: 229-494-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR