2,5-Dichlorbenzophenon
Lieferant: FLUOROCHEM
Synonyme:
(2,5-Dichlorophenyl)(phenyl)methanone
018517-10G
FLUO018517-10GEA
91.4
EUR
FLUO018517-10G
2,5-Dichlorbenzophenon
2,5-Dichlorbenzophenon
Formel:
C₁₃H₈Cl₂O Molecular Weight: 251,11 g/mol |
MDL:
MFCD00079746 CAS-Nummer: 16611-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...