3,4-Dichlorbenzylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1,2-Dichlor-4-(isothiocyanatomethyl)benzol

018522-5G
FLUO018522-5GEA 306 EUR
FLUO018522-5G
3,4-Dichlorbenzylisothiocyanat
3,4-Dichlorbenzylisothiocyanat
Formel: C₈H₅Cl₂NS
Molecular Weight: 218.11 g/mol
MDL: MFCD00041113
CAS-Nummer: 18967-42-5
UN: 2923
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR