4,6-Dichlorsalicylaldehyd

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dichlor-6-hydroxybenzaldehyd

018525-1G
FLUO018525-1GEA 237 EUR
FLUO018525-1G
4,6-Dichlorsalicylaldehyd
4,6-Dichlorsalicylaldehyd
Formel: C₇H₄Cl₂O₂
Molecular Weight: 191,01 g/mol
MDL: MFCD00016590
CAS-Nummer: 78443-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR