2,5-Dichlor-3-nitrobenzoesäure
Lieferant: FLUOROCHEM
|
Achtung
|
018529-1G
FLUO018529-1GEA
110
EUR
FLUO018529-1G
2,5-Dichlor-3-nitrobenzoesäure
2,5-Dichloro-3-nitrobenzoic acid
Formel:
C₇H₃Cl₂NO₄ |
MDL:
MFCD00014691 CAS-Nummer: 88-86-8 EINECS: 201-862-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...