2-(2,4-Dichlorphenoxy)ethanol

Lieferant: FLUOROCHEM

018531-10G
FLUO018531-10GEA 260 EUR
FLUO018531-10G
2-(2,4-Dichlorphenoxy)ethanol
2-(2,4-Dichlorphenoxy)ethanol
Formel: C₈H₈Cl₂O₂
Molecular Weight: 207.06 g/mol
MDL: MFCD00016564
CAS-Nummer: 120-67-2
EINECS: 204-416-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR