3,5-Dichlorphenylhydraziniumchlorid
Lieferant: FLUOROCHEM
018533-10G
FLUO018533-10GEA
51.9
EUR
FLUO018533-10G
3,5-Dichlorphenylhydraziniumchlorid
3,5-Dichlorphenylhydraziniumchlorid
Formel:
C₆H₇Cl₃N₂ |
MDL:
MFCD00012938 CAS-Nummer: 63352-99-8 EINECS: 264-109-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...