3-(2,4-Dichlorphenyl)propionsäure

Lieferant: FLUOROCHEM

018534-10G
FLUO018534-10GEA 152 EUR
FLUO018534-10G
3-(2,4-Dichlorphenyl)propionsäure
3-(2,4-Dichlorphenyl)propionsäure
Formel: C₉H₈Cl₂O₂
MDL: MFCD00275605
CAS-Nummer: 55144-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR