3-(3,4-Dichlorphenyl)propionsäure

Lieferant: FLUOROCHEM

Synonyme: 3,4-Dichlorhydrozimtsäure

018536-5G
FLUO018536-5GEA 135 EUR
FLUO018536-5G
3-(3,4-Dichlorphenyl)propionsäure
3-(3,4-Dichlorphenyl)propionsäure
Formel: C₉H₈Cl₂O₂
Molecular Weight: 219.07 g/mol
MDL: MFCD00016551
CAS-Nummer: 25173-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR