4-(2,4-Dichlorphenyl)-3-thiosemicarbazid
Lieferant: FLUOROCHEM
018538-2G
FLUO018538-2GEA
0
EUR
FLUO018538-2G
4-(2,4-Dichlorphenyl)-3-thiosemicarbazid
4-(2,4-Dichlorphenyl)-3-thiosemicarbazid
Formel:
C₇H₇Cl₂N₃S Molecular Weight: 236,12 g/mol |
MDL:
MFCD00041272 CAS-Nummer: 13124-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...