4-(2,5-Dichlorphenyl)-3-thiosemicarbazid

Lieferant: FLUOROCHEM

018539-1G
FLUO018539-1GEA 39.9 EUR
FLUO018539-1G
4-(2,5-Dichlorphenyl)-3-thiosemicarbazid
4-(2,5-Dichlorphenyl)-3-thiosemicarbazid
Formel: C₇H₇Cl₂N₃S
MDL: MFCD00041273
CAS-Nummer: 14580-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR