4-(3,4-Dichlorphenyl)-3-thiosemicarbazid
Lieferant: FLUOROCHEM
018541-2G
FLUO018541-2GEA
76.2
EUR
FLUO018541-2G
4-(3,4-Dichlorphenyl)-3-thiosemicarbazid
4-(3,4-Dichlorphenyl)-3-thiosemicarbazid
Formel:
C₇H₇Cl₂N₃S |
MDL:
MFCD00041288 CAS-Nummer: 13124-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...