1-(2,3-Dichlorphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
018542-5G
FLUO018542-5GEA
230
EUR
FLUO018542-5G
1-(2,3-Dichlorphenyl)-2-thioharnstoff
1-(2,3-Dichlorphenyl)-2-thioharnstoff
Formel:
(C₆H₃Cl₂)NC(S)NH₂ Molecular Weight: 221,11 g/mol |
MDL:
MFCD00060428 CAS-Nummer: 41542-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...