1-(2,5-Dichlorphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
Synonyme:
N-(2,5-Dichlorphenyl)thioharnstoff, 2,5-Dichlorphenylthioharnstoff
018544-5G
FLUO018544-5GEA
186
EUR
FLUO018544-5G
1-(2,5-Dichlorphenyl)-2-thioharnstoff
1-(2,5-Dichlorphenyl)-2-thioharnstoff
Formel:
(C₆H₃Cl₂)NC(S)NH₂ Molecular Weight: 221,11 g/mol Schmelzpunkt: 320 °C |
MDL:
MFCD00041154 CAS-Nummer: 4949-85-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...