1-(3,4-Dichlorphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
N-(3,4-Dichlorphenyl)thioharnstoff, 3,4-Dichlorphenylthioharnstoff
018546-10G
FLUO018546-10GEA
112
EUR
FLUO018546-10G
1-(3,4-Dichlorphenyl)-2-thioharnstoff
1-(3,4-Dichlorphenyl)-2-thioharnstoff
Formel:
(C₆H₃Cl₂)NC(S)NH₂ Molecular Weight: 221,11 g/mol Siedepunkt: 326 °C (1013 hPa) Schmelzpunkt: 220 °C |
MDL:
MFCD00004935 CAS-Nummer: 19250-09-0 EINECS: 242-919-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...