1-(3,5-Dichlorphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
N-(3,5-Dichlorphenyl)thioharnstoff, 3,5-Dichlorphenylthioharnstoff
018547-1G
FLUO018547-1GEA
134
EUR
FLUO018547-1G
1-(3,5-Dichlorphenyl)-2-thioharnstoff
1-(3,5-Dichlorphenyl)-2-thioharnstoff
Formel:
(C₆H₃Cl₂)NC(S)NH₂ Molecular Weight: 221,11 g/mol Siedepunkt: 326 °C (1013 hPa) Schmelzpunkt: 180 °C |
MDL:
MFCD00041168 CAS-Nummer: 107707-33-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...