1,3-Dicyclohexyl-2-thioharnstoff

Lieferant: FLUOROCHEM

018550-5G
FLUO018550-5GEA 48.8 EUR
FLUO018550-5G
1,3-Dicyclohexyl-2-thioharnstoff
1,3-Dicyclohexyl-2-thioharnstoff
Formel: C₁₃H₂₄N₂S
MDL: MFCD00021316
CAS-Nummer: 1212-29-9
EINECS: 214-920-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR